Product Details

SNP ID
rs142526170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:124360654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTCCTAACCAAAAGATCTCTATT[A/G]ACCCAAACTTGTGCCCAAGAATAGC
Phenotype
MIM: 300490
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SH2D1A PubMed Links

Gene Details

Gene
SH2D1A
Gene Name
SH2 domain containing 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114937.2 Intron NP_001108409.1
NM_002351.4 Intron NP_002342.1

View Full Product Details