Product Details

SNP ID
rs142772459
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:15321540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGATATCTCATTATTCTCACCC[A/C]CTCTTTTACTGTGAGAATAGTTATT
Phenotype
MIM: 300626 MIM: 311770
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ASB11 PubMed Links

Gene Details

Gene
ASB11
Gene Name
ankyrin repeat and SOCS box containing 11
There are no transcripts associated with this gene.

Gene
PIGA
Gene Name
phosphatidylinositol glycan anchor biosynthesis class A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002641.3 2361 Intron NP_002632.1
NM_020473.3 2361 Intron NP_065206.3
XM_011545539.2 2361 Missense Mutation GGG,GTG G243V XP_011543841.1
XM_017029581.1 2361 Intron XP_016885070.1

View Full Product Details