Product Details

SNP ID
rs143422691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154652405 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGTTTGGGTGTTCTGAGATCTC[C/T]AGCTTTCTGCCCCTCCGGGGAGGCG
Phenotype
MIM: 300396
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTAG2 PubMed Links

Gene Details

Gene
CTAG2
Gene Name
cancer/testis antigen 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020994.4 Intron NP_066274.2
NM_172377.4 Intron NP_758965.2

View Full Product Details