Product Details
- SNP ID
-
rs143631197
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:2221186 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCACCAACTCCTTCCAGCTCTGGG[G/T]TGACTGCTGCGTAGATGGAAGTCCA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PubMed Links
Gene Details
View Full Product Details