Product Details

SNP ID
rs144174608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:53381934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGAAGGGGTGAAAGGTGATGAT[C/T]AAGCTAGATTTACATGGAAAGGGAT
Phenotype
MIM: 300040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMC1A PubMed Links

Gene Details

Gene
SMC1A
Gene Name
structural maintenance of chromosomes 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281463.1 Intron NP_001268392.1
NM_006306.3 Intron NP_006297.2

View Full Product Details