Product Details

SNP ID
rs144346983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:54809901 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGGTCTCAAAGACCCCAGAGGC[C/T]CGGGAGGCACCTGCCACCCAGGCCT
Phenotype
MIM: 300470 MIM: 300662
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAGED2 PubMed Links

Gene Details

Gene
MAGED2
Gene Name
MAGE family member D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014599.5 418 Silent Mutation GCC,GCT A75A NP_055414.2
NM_177433.2 418 Silent Mutation GCC,GCT A75A NP_803182.1
NM_201222.2 418 Silent Mutation GCC,GCT A75A NP_957516.1
Gene
SNORA11
Gene Name
small nucleolar RNA, H/ACA box 11
There are no transcripts associated with this gene.

View Full Product Details