Product Details

SNP ID
rs144492438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:136875511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCCATCATCTCTTGGGGACAAAT[A/G]AACATCTCTACGAGAGGGCAGCGGT
Phenotype
MIM: 300199
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RBMX PubMed Links

Gene Details

Gene
RBMX
Gene Name
RNA binding motif protein, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164803.1 Intron NP_001158275.1
NM_002139.3 Intron NP_002130.2
Gene
SNORD61
Gene Name
small nucleolar RNA, C/D box 61
There are no transcripts associated with this gene.

View Full Product Details