Product Details

SNP ID
rs145056272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:149482778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAACAACTGGAAAAGATCTCCAC[A/C]TTGGGAATCATTATACATATTGTGA
Phenotype
MIM: 300823
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IDS PubMed Links

Gene Details

Gene
IDS
Gene Name
iduronate 2-sulfatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000202.7 1794 Missense Mutation GGT,TGT G541C NP_000193.1
NM_001166550.3 1794 Missense Mutation GGT,TGT G451C NP_001160022.1
NM_006123.4 1794 Intron NP_006114.1

View Full Product Details