Product Details

SNP ID
rs145546830
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47221135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTAGCGTGTATTAATGGGGATC[A/G]GGACATAGGGTGCGATGACCAAGTC
Phenotype
MIM: 311550 MIM: 314370
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDK16 PubMed Links

Gene Details

Gene
CDK16
Gene Name
cyclin dependent kinase 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170460.1 227 Intron NP_001163931.1
NM_006201.4 227 Intron NP_006192.1
NM_033018.3 227 Intron NP_148978.2
XM_011543920.2 227 Intron XP_011542222.1
XM_011543921.1 227 Intron XP_011542223.1
XM_011543922.1 227 Intron XP_011542224.1
XM_011543923.1 227 Intron XP_011542225.1
XM_011543924.1 227 Intron XP_011542226.1
XM_011543925.2 227 Intron XP_011542227.1
XM_011543926.1 227 UTR 5 XP_011542228.1
XM_011543927.2 227 Intron XP_011542229.1
XM_011543928.1 227 Intron XP_011542230.1
XM_017029569.1 227 Intron XP_016885058.1
XM_017029570.1 227 Intron XP_016885059.1
XM_017029571.1 227 UTR 5 XP_016885060.1
XM_017029572.1 227 Intron XP_016885061.1
XM_017029573.1 227 Intron XP_016885062.1
Gene
UBA1
Gene Name
ubiquitin like modifier activating enzyme 1
There are no transcripts associated with this gene.

View Full Product Details