Product Details

SNP ID
rs146439706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:102127229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAAAGCGCCCAGCTGAGGATGA[C/T]ATACCCAGGAAAGCCAAAAGAAAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TCEAL2 PubMed Links

Gene Details

Gene
TCEAL2
Gene Name
transcription elongation factor A like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080390.3 618 Silent Mutation GAC,GAT D133D NP_525129.1

View Full Product Details