Product Details

SNP ID
rs146482073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49831644 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACTTAAAATGTAAGGAAAAGTT[A/G]TCCAAGTACAGTGAACTCCTGTGTA
Phenotype
MIM: 300287
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAGE4 PubMed Links

Gene Details

Gene
PAGE4
Gene Name
PAGE family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318877.1 Intron NP_001305806.1
NM_007003.4 Intron NP_008934.1

View Full Product Details