Product Details

SNP ID
rs146919548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:78755069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCTCTGTTTGTCTTCTGTTTCC[A/G]CATGAAAATGAGAAGTGAGACTGCT
Phenotype
MIM: 300086
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LPAR4 PubMed Links

Gene Details

Gene
LPAR4
Gene Name
lysophosphatidic acid receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278000.1 605 Missense Mutation CAC,CGC H67R NP_001264929.1
NM_005296.2 605 Missense Mutation CAC,CGC H67R NP_005287.1
XM_005262126.3 605 Missense Mutation CAC,CGC H67R XP_005262183.1
XM_006724639.3 605 Missense Mutation CAC,CGC H67R XP_006724702.1
XM_017029437.1 605 Missense Mutation CAC,CGC H67R XP_016884926.1
XM_017029438.1 605 Missense Mutation CAC,CGC H67R XP_016884927.1

View Full Product Details