Product Details

SNP ID
rs147712053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:79363133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAGCACATCTCTCCACGGTAAA[C/T]GGTGCTCTAAAAGACAAAAAGGGAA
Phenotype
MIM: 300222
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITM2A PubMed Links

Gene Details

Gene
ITM2A
Gene Name
integral membrane protein 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171581.1 587 Missense Mutation ATT,GTT I40V NP_001165052.1
NM_004867.4 587 Missense Mutation ATT,GTT I84V NP_004858.1

View Full Product Details