Product Details

SNP ID
rs147731515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:50281273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGGCCAATCACCACTGCACTTC[A/T]TGACGGCTGTAGTTTTCAAAATCCC
Phenotype
MIM: 300456
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCNB3 PubMed Links

Gene Details

Gene
CCNB3
Gene Name
cyclin B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033031.2 Intron NP_149020.2
NM_033670.2 Intron NP_391990.1
XM_017029913.1 Intron XP_016885402.1
XM_017029914.1 Intron XP_016885403.1
XM_017029915.1 Intron XP_016885404.1
XM_017029916.1 Intron XP_016885405.1
XM_017029917.1 Intron XP_016885406.1
XM_017029918.1 Intron XP_016885407.1

View Full Product Details