Product Details

SNP ID
hCV175549821
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:152728103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGTGTTGATGGGTGGTTGTTGGA[C/A]AATGGGCTGGAGGCTCGTGGTTTCC
Phenotype
MIM: 300944 MIM: 300177 MIM: 300549
Polymorphism
C/A, Transversion Substitution
Allele Nomenclature
Literature Links
CSAG1 PubMed Links

Gene Details

Gene
CSAG1
Gene Name
chondrosarcoma associated gene 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102576.2 467 Intron NP_001096046.2
NM_153478.2 467 Missense Mutation TTG,TTT L46F NP_705611.2
Gene
CSAG4
Gene Name
CSAG family member 4 (pseudogene)
There are no transcripts associated with this gene.

Gene
MAGEA12
Gene Name
MAGE family member A12
There are no transcripts associated with this gene.

Gene
MAGEA2B
Gene Name
MAGE family member A2B
There are no transcripts associated with this gene.

View Full Product Details