Product Details
- SNP ID
-
rs148094474
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:68839673 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCCTCTTCCTGCAGCAACATCCA[A/T]TGGAAGCCTGGAGGGGCTGGAAAAC
- Phenotype
-
MIM: 300035
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
EFNB1
PubMed Links
Gene Details
- Gene
- EFNB1
- Gene Name
- ephrin B1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004429.4 |
1196 |
Missense Mutation |
AAT,ATT |
N139I |
NP_004420.1 |
View Full Product Details