Product Details

SNP ID
rs149174700
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:21839760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATTCCGGTGTCGCTGGTGGTGG[G/T]GGTGGTGGGTGGCTGGACTGTCGTC
Phenotype
MIM: 300294
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MBTPS2 PubMed Links

Gene Details

Gene
MBTPS2
Gene Name
membrane bound transcription factor peptidase, site 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015884.3 223 Missense Mutation GGG,GTG G9V NP_056968.1

View Full Product Details