Product Details
- SNP ID
-
rs149475033
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:17753317 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGACGTGTTGCTGAAGCACTTGGG[A/G]GTGAAGCTGGGAACGGCTGTGAAGC
- Phenotype
-
MIM: 300227
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SCML1
PubMed Links
Gene Details
- Gene
- SCML1
- Gene Name
- sex comb on midleg-like 1 (Drosophila)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001037535.2 |
4082 |
Silent Mutation |
GGA,GGG |
G184G |
NP_001032624.1 |
NM_001037536.2 |
4082 |
Silent Mutation |
GGA,GGG |
G184G |
NP_001032625.1 |
NM_001037540.2 |
4082 |
Silent Mutation |
GGA,GGG |
G305G |
NP_001032629.1 |
NM_006746.5 |
4082 |
Silent Mutation |
GGA,GGG |
G278G |
NP_006737.2 |
XM_005274578.2 |
4082 |
Silent Mutation |
GGA,GGG |
G306G |
XP_005274635.1 |
XM_005274579.2 |
4082 |
Silent Mutation |
GGA,GGG |
G277G |
XP_005274636.1 |
XM_006724508.3 |
4082 |
Silent Mutation |
GGA,GGG |
G305G |
XP_006724571.1 |
XM_006724509.3 |
4082 |
Silent Mutation |
GGA,GGG |
G277G |
XP_006724572.1 |
XM_011545564.2 |
4082 |
Silent Mutation |
GGA,GGG |
G306G |
XP_011543866.1 |
XM_017029723.1 |
4082 |
Silent Mutation |
GGA,GGG |
G278G |
XP_016885212.1 |
View Full Product Details