Product Details

SNP ID
rs150070913
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:102937375 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGACTTCCTGCTCAAGTTCCTGCT[G/T]GTGGGCGACAGGGACGTAGGCAAGA
Phenotype
MIM: 300405
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RAB40AL PubMed Links

Gene Details

Gene
RAB40AL
Gene Name
RAB40A, member RAS oncogene family-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031834.1 104 Silent Mutation CTG,CTT L19L NP_001027004.1

View Full Product Details