Product Details

SNP ID
rs150079861
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101616590 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCCCAATCATCAGTCCTGCCG[A/C]CATCCAACCCACTTCCCGAGCCCGG
Phenotype
MIM: 300364
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARMCX3 PubMed Links

Gene Details

Gene
ARMCX3
Gene Name
armadillo repeat containing, X-linked 3
There are no transcripts associated with this gene.

Gene
ARMCX6
Gene Name
armadillo repeat containing, X-linked 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009584.1 464 Missense Mutation GCG,TCG A11S NP_001009584.1
NM_001184768.1 464 Missense Mutation GCG,TCG A11S NP_001171697.1
NM_019007.3 464 Missense Mutation GCG,TCG A11S NP_061880.2
XM_011530976.2 464 Missense Mutation GCG,TCG A74S XP_011529278.1

View Full Product Details