Product Details

SNP ID
rs150788310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48481335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTGAAGCCAGGGGGCTGCTTTG[C/T]GGCCAAGGTAAGTCTCAAGGAACTT
Phenotype
MIM: 300499
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FTSJ1 PubMed Links

Gene Details

Gene
FTSJ1
Gene Name
FtsJ RNA methyltransferase homolog 1 (E. coli)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282157.1 879 Missense Mutation GCG,GTG A17V NP_001269086.1
NM_012280.3 879 Missense Mutation GCG,GTG A154V NP_036412.1
NM_177439.2 879 Missense Mutation GCG,GTG A154V NP_803188.1
XM_005272595.1 879 Missense Mutation GCG,GTG A154V XP_005272652.1
XM_011543893.2 879 Missense Mutation GCG,GTG A154V XP_011542195.1
XM_011543894.2 879 Missense Mutation GCG,GTG A154V XP_011542196.1
XM_011543895.1 879 Missense Mutation GCG,GTG A154V XP_011542197.1
XM_017029382.1 879 Missense Mutation GCG,GTG A154V XP_016884871.1

View Full Product Details