Product Details
- SNP ID
-
rs150826975
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:19929870 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAACCGTAAACGCTGATACTTAGCC[C/T]GAATGTACAAGCTCTTCTGTACCAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CXorf23
PubMed Links
Gene Details
- Gene
- CXorf23
- Gene Name
- chromosome X open reading frame 23
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_198279.3 |
2188 |
Missense Mutation |
CAG,CGG |
Q645R |
NP_938020.2 |
XM_005274473.3 |
2188 |
Missense Mutation |
CAG,CGG |
Q674R |
XP_005274530.1 |
XM_005274474.3 |
2188 |
Missense Mutation |
CAG,CGG |
Q654R |
XP_005274531.1 |
XM_005274475.3 |
2188 |
Missense Mutation |
CAG,CGG |
Q644R |
XP_005274532.1 |
XM_011545475.2 |
2188 |
Missense Mutation |
CAG,CGG |
Q674R |
XP_011543777.1 |
XM_011545476.2 |
2188 |
Missense Mutation |
CAG,CGG |
Q674R |
XP_011543778.1 |
XM_011545477.2 |
2188 |
Missense Mutation |
CAG,CGG |
Q645R |
XP_011543779.1 |
XM_011545478.2 |
2188 |
Intron |
|
|
XP_011543780.1 |
XM_011545479.2 |
2188 |
Missense Mutation |
CAG,CGG |
Q615R |
XP_011543781.1 |
XM_011545482.2 |
2188 |
Intron |
|
|
XP_011543784.1 |
XM_017029390.1 |
2188 |
Missense Mutation |
CAG,CGG |
Q625R |
XP_016884879.1 |
XM_017029391.1 |
2188 |
Missense Mutation |
CAG,CGG |
Q624R |
XP_016884880.1 |
XM_017029392.1 |
2188 |
Intron |
|
|
XP_016884881.1 |
View Full Product Details