Product Details

SNP ID
rs151072020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:57910186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGTCGCCGCCGCCGCTAGGTTGAT[C/G]GGGCCTCGGCGCGAACAGGCTTGGG
Phenotype
MIM: 300235
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZXDA PubMed Links

Gene Details

Gene
ZXDA
Gene Name
zinc finger, X-linked, duplicated A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007156.4 448 Missense Mutation CAT,GAT H79D NP_009087.1

View Full Product Details