Product Details

SNP ID
rs11544587
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132370313 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCAGCTGGAGCAGGAGAGCTGTC[C/T]GGATGGCTGGGAGTTCAGTCAGGAC
Phenotype
MIM: 603377
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC553103 PubMed Links

Gene Details

Gene
LOC553103
Gene Name
uncharacterized LOC553103
There are no transcripts associated with this gene.

Gene
MIR3936
Gene Name
microRNA 3936
There are no transcripts associated with this gene.

Gene
SLC22A5
Gene Name
solute carrier family 22 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308122.1 182 Missense Mutation CCG,CTG P114L NP_001295051.1
NM_003060.3 182 Missense Mutation CCG,CTG P114L NP_003051.1
XM_011543590.2 182 UTR 5 XP_011541892.1
XM_017009778.1 182 UTR 5 XP_016865267.1

View Full Product Details