Product Details

SNP ID
rs5078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:116836061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCGCTGTAGGGGGCCAGATGCGTG[G/C]GCAGCGCGTCCACATGGGCGCGCGC
Phenotype
MIM: 107680 MIM: 107720 MIM: 614776
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
APOA1 PubMed Links

Gene Details

Gene
APOA1
Gene Name
apolipoprotein A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000039.2 622 Missense Mutation CCC,CGC P184R NP_000030.1
NM_001318017.1 622 Missense Mutation CCC,CGC P184R NP_001304946.1
NM_001318018.1 622 Missense Mutation CCC,CGC P184R NP_001304947.1
NM_001318021.1 622 Missense Mutation CCC,CGC P75R NP_001304950.1
Gene
APOA1-AS
Gene Name
APOA1 antisense RNA
There are no transcripts associated with this gene.

Gene
APOC3
Gene Name
apolipoprotein C3
There are no transcripts associated with this gene.

Gene
SIK3
Gene Name
SIK family kinase 3
There are no transcripts associated with this gene.

View Full Product Details