Product Details

SNP ID
rs1058444
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84122675 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTATTTCCCACATTCTTAATGA[C/G]ACCCCATCCACAGTGGTCCAATCAA
Phenotype
MIM: 603355
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HSDL1 PubMed Links

Gene Details

Gene
HSDL1
Gene Name
hydroxysteroid dehydrogenase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146051.1 2967 UTR 3 NP_001139523.1
NM_031463.4 2967 UTR 3 NP_113651.4
XM_005256189.2 2967 UTR 3 XP_005256246.1
Gene
LOC729887
Gene Name
uncharacterized LOC729887
There are no transcripts associated with this gene.

Gene
MBTPS1
Gene Name
membrane bound transcription factor peptidase, site 1
There are no transcripts associated with this gene.

View Full Product Details