Product Details
- SNP ID
-
rs11150816
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:79936215 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCGTGGCAGGAGTGGGCAGAATGGG[A/G]TGCATTTTACAGAAGAGTCACAGAG
- Phenotype
-
MIM: 616637
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101928766
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3169598] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101928766
- Gene Name
- uncharacterized LOC101928766
There are no transcripts associated with this gene.
- Gene
- TBC1D16
- Gene Name
- TBC1 domain family member 16
View Full Product Details