Product Details
- SNP ID
-
rs11189222
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:97441820 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTTTTTTTTTTTTAAATCTTTTC[A/C]CAACTTGCCCTTCAGAGAGGTATAG
- Phenotype
-
MIM: 606493
MIM: 172250
MIM: 616750
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
EXOSC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12784013] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EXOSC1
- Gene Name
- exosome component 1
- Gene
- PGAM1
- Gene Name
- phosphoglycerate mutase 1
There are no transcripts associated with this gene.
- Gene
- ZDHHC16
- Gene Name
- zinc finger DHHC-type containing 16
There are no transcripts associated with this gene.
View Full Product Details