Product Details

SNP ID
rs11188846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:96599118 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCACTAGCAGATGGAAAGGAGGG[C/T]GGATCTGACAGCTGAAAGAGCAGCA
Phenotype
MIM: 607942
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PIK3AP1 PubMed Links

Gene Details

Gene
PIK3AP1
Gene Name
phosphoinositide-3-kinase adaptor protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152309.2 Intron NP_689522.2
XM_005269498.1 Intron XP_005269555.1
XM_005269499.1 Intron XP_005269556.1
XM_011539248.1 Intron XP_011537550.1
XM_011539249.1 Intron XP_011537551.1

View Full Product Details