Product Details

SNP ID
rs11195253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:110650901 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATCTCCTCCTTTGGTTTATTTT[C/G]AATTAATTATTGAACAGAATGATTT
Phenotype
MIM: 613171
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RBM20 PubMed Links

Gene Details

Gene
RBM20
Gene Name
RNA binding motif protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134363.2 Intron NP_001127835.2
XM_011539697.2 Intron XP_011537999.1
XM_017016103.1 Intron XP_016871592.1
XM_017016104.1 Intron XP_016871593.1

View Full Product Details