Product Details

SNP ID
rs11541292
Assay Type
Functionally Tested
NCBI dbSNP Submissions
5
Location
Chr.1:6591800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCACAGGGCAAGCAACAGTGCC[G/A]GAGGGTGCCTGGTCACAGGAAGCCC
Phenotype
MIM: 616262 MIM: 165270
Polymorphism
G/A, Transition Substitution
Allele Nomenclature
Literature Links
KLHL21 PubMed Links

Gene Details

Gene
KLHL21
Gene Name
kelch like family member 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324309.1 3440 UTR 3 NP_001311238.1
NM_014851.3 3440 UTR 3 NP_055666.2
Gene
ZBTB48
Gene Name
zinc finger and BTB domain containing 48
There are no transcripts associated with this gene.

View Full Product Details