Product Details
- SNP ID
-
rs11539850
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:37679631 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCATCACCTTCGACCAGGCCAACC[C/T]GACCGTCAAGCTGCCAGATGGATAC
- Phenotype
-
MIM: 150570
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LGALS1
PubMed Links
Gene Details
- Gene
- LGALS1
- Gene Name
- galectin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002305.3 |
387 |
Missense Mutation |
CCG,CTG |
P97L |
NP_002296.1 |
- Gene
- NOL12
- Gene Name
- nucleolar protein 12
There are no transcripts associated with this gene.
View Full Product Details