Product Details

SNP ID
rs11545807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137205129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGGCACAACAGCATCTCGAAGA[C/A]GAACTTGAGACTCAGGACCGTAAGT
Phenotype
MIM: 606073 MIM: 616499 MIM: 613354
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
NDOR1 PubMed Links

Gene Details

Gene
NDOR1
Gene Name
NADPH dependent diflavin oxidoreductase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144026.2 510 Intron NP_001137498.1
NM_001144027.2 510 Intron NP_001137499.1
NM_001144028.2 510 Intron NP_001137500.1
NM_014434.3 510 Intron NP_055249.1
XM_011518545.1 510 Intron XP_011516847.1
XM_011518547.1 510 Intron XP_011516849.1
XM_011518548.2 510 Intron XP_011516850.1
XM_017014626.1 510 Intron XP_016870115.1
XM_017014627.1 510 Intron XP_016870116.1
Gene
TMEM203
Gene Name
transmembrane protein 203
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053045.1 510 Missense Mutation GTC,TTC V96F NP_444273.1
Gene
TPRN
Gene Name
taperin
There are no transcripts associated with this gene.

View Full Product Details