Product Details
- SNP ID
-
rs11546629
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:49129522 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCAATGACTGTGGGTTCCAAGTC[T/A]ACAAACACAGCCCGGGGCACGTGCT
- Phenotype
-
MIM: 602530
- Polymorphism
- T/A, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TUBA1B
PubMed Links
Gene Details
- Gene
- TUBA1B
- Gene Name
- tubulin alpha 1b
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006082.2 |
425 |
Silent Mutation |
GTA,GTT |
V68V |
NP_006073.2 |
View Full Product Details