Product Details

SNP ID
rs11547532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:102082325 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTTTCTTTGCTGCCATGTAACC[T/C]ATTGTTGAGTTGTCTCTTAGGGCTT
Phenotype
MIM: 140571
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
HSP90AA1 PubMed Links

Gene Details

Gene
HSP90AA1
Gene Name
heat shock protein 90 alpha family class A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017963.2 2065 Missense Mutation ATA,ATG I747M NP_001017963.2
NM_005348.3 2065 Missense Mutation ATA,ATG I625M NP_005339.3
XM_011536718.2 2065 Missense Mutation ATA,ATG I746M XP_011535020.1

View Full Product Details