Product Details

SNP ID
rs11551120
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44476222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGGAATTGCCAGGTTGAAAAGTC[A/G]TTCATAGACTTTCCCCCTTTCTTTT
Phenotype
MIM: 606740
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF180 PubMed Links

Gene Details

Gene
ZNF180
Gene Name
zinc finger protein 180
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278508.2 2420 UTR 3 NP_001265437.2
NM_001278509.2 2420 UTR 3 NP_001265438.2
NM_001288759.2 2420 UTR 3 NP_001275688.2
NM_001288760.2 2420 UTR 3 NP_001275689.1
NM_001288761.2 2420 UTR 3 NP_001275690.1
NM_001288762.2 2420 UTR 3 NP_001275691.1
NM_001291633.1 2420 UTR 3 NP_001278562.1
NM_013256.5 2420 UTR 3 NP_037388.3
XM_011527280.2 2420 UTR 3 XP_011525582.1
XM_017027250.1 2420 UTR 3 XP_016882739.1
XM_017027251.1 2420 UTR 3 XP_016882740.1
XM_017027252.1 2420 UTR 3 XP_016882741.1
XM_017027253.1 2420 UTR 3 XP_016882742.1
XM_017027254.1 2420 UTR 3 XP_016882743.1
XM_017027255.1 2420 UTR 3 XP_016882744.1
XM_017027256.1 2420 UTR 3 XP_016882745.1

View Full Product Details