Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_138383.2 | 4315 | UTR 3 | NP_612392.1 | ||
XM_005256236.4 | 4315 | UTR 3 | XP_005256293.2 | ||
XM_011523450.2 | 4315 | UTR 3 | XP_011521752.1 | ||
XM_011523453.2 | 4315 | UTR 3 | XP_011521755.2 | ||
XM_011523454.2 | 4315 | UTR 3 | XP_011521756.2 | ||
XM_011523458.2 | 4315 | UTR 3 | XP_011521760.1 | ||
XM_017023879.1 | 4315 | UTR 3 | XP_016879368.1 | ||
XM_017023880.1 | 4315 | UTR 3 | XP_016879369.1 | ||
XM_017023881.1 | 4315 | UTR 3 | XP_016879370.1 | ||
XM_017023882.1 | 4315 | Intron | XP_016879371.1 | ||
XM_017023883.1 | 4315 | Intron | XP_016879372.1 | ||
XM_017023884.1 | 4315 | Intron | XP_016879373.1 |