Product Details

SNP ID
rs174690
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19951909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGAGACAGGGGCCCCATCTTCA[A/G]GTGTTGGCCAGAACACAGGAAATTC
Phenotype
MIM: 116790 MIM: 606448
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COMT PubMed Links

Gene Details

Gene
COMT
Gene Name
catechol-O-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000754.3 Intron NP_000745.1
NM_001135161.1 Intron NP_001128633.1
NM_001135162.1 Intron NP_001128634.1
NM_007310.2 Intron NP_009294.1
XM_011529886.1 Intron XP_011528188.1
XM_017028594.1 Intron XP_016884083.1
XM_017028595.1 Intron XP_016884084.1
Gene
TXNRD2
Gene Name
thioredoxin reductase 2
There are no transcripts associated with this gene.

View Full Product Details