Product Details

SNP ID
rs180781103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:38750105 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGCTTCTTTTTGAGTGCTACGG[A/G]ATTCCCAAGGTTGCCTATGGAATAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTR5 PubMed Links

Gene Details

Gene
ACTR5
Gene Name
ARP5 actin-related protein 5 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024855.3 496 Silent Mutation GGA,GGG G157G NP_079131.3

View Full Product Details