Product Details
- SNP ID
-
rs180907536
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:85979723 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGGGGGGTTATCAGGAATGTCAA[A/G]TGACTCCTCTTCAAAATCATCATCC
- Phenotype
-
MIM: 607385
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MGAT4C
PubMed Links
Gene Details
- Gene
- MGAT4C
- Gene Name
- MGAT4 family member C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_013244.3 |
1335 |
Missense Mutation |
CTT,TTT |
L335F |
NP_037376.2 |
XM_005268776.4 |
1335 |
Missense Mutation |
CTT,TTT |
L364F |
XP_005268833.1 |
XM_005268779.3 |
1335 |
Missense Mutation |
CTT,TTT |
L335F |
XP_005268836.1 |
XM_011538150.2 |
1335 |
Missense Mutation |
CTT,TTT |
L373F |
XP_011536452.1 |
XM_011538151.2 |
1335 |
Missense Mutation |
CTT,TTT |
L373F |
XP_011536453.1 |
XM_011538156.2 |
1335 |
Missense Mutation |
CTT,TTT |
L335F |
XP_011536458.1 |
XM_011538157.2 |
1335 |
Missense Mutation |
CTT,TTT |
L335F |
XP_011536459.1 |
XM_017019143.1 |
1335 |
Missense Mutation |
CTT,TTT |
L384F |
XP_016874632.1 |
XM_017019144.1 |
1335 |
Missense Mutation |
CTT,TTT |
L373F |
XP_016874633.1 |
XM_017019145.1 |
1335 |
Missense Mutation |
CTT,TTT |
L364F |
XP_016874634.1 |
XM_017019146.1 |
1335 |
Missense Mutation |
CTT,TTT |
L364F |
XP_016874635.1 |
XM_017019147.1 |
1335 |
Missense Mutation |
CTT,TTT |
L364F |
XP_016874636.1 |
XM_017019148.1 |
1335 |
Missense Mutation |
CTT,TTT |
L335F |
XP_016874637.1 |
XM_017019149.1 |
1335 |
Missense Mutation |
CTT,TTT |
L335F |
XP_016874638.1 |
XM_017019150.1 |
1335 |
Missense Mutation |
CTT,TTT |
L335F |
XP_016874639.1 |
View Full Product Details