Product Details

SNP ID
rs180946601
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:167094860 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTGGCAGGTTGTCATGGCGACCA[A/G]AAAGGACACAGAGGAGGAGCAGGTA
Phenotype
MIM: 602171
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP27 PubMed Links

Gene Details

Gene
DUSP27
Gene Name
dual specificity phosphatase 27 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080426.1 174 Missense Mutation AAA,AGA K4R NP_001073895.1
XM_005245603.3 174 Missense Mutation AAA,AGA K4R XP_005245660.1
XM_011510146.2 174 Intron XP_011508448.1
Gene
GPA33
Gene Name
glycoprotein A33
There are no transcripts associated with this gene.

Gene
LOC105371598
Gene Name
uncharacterized LOC105371598
There are no transcripts associated with this gene.

View Full Product Details