Product Details

SNP ID
rs180999314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:54308671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTATTTCACTAGGAATCACACC[A/G]TTTTGCAAGGTACTCGATTCAAAAG
Phenotype
MIM: 606749
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TINAG PubMed Links

Gene Details

Gene
TINAG
Gene Name
tubulointerstitial nephritis antigen
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014464.3 267 Missense Mutation ATT,GTT I41V NP_055279.3
XM_006715062.2 267 Missense Mutation ATT,GTT I23V XP_006715125.1
XM_011514497.2 267 Missense Mutation ATT,GTT I41V XP_011512799.1
XM_017010745.1 267 Missense Mutation ATT,GTT I41V XP_016866234.1
XM_017010746.1 267 Missense Mutation ATT,GTT I41V XP_016866235.1
XM_017010747.1 267 Missense Mutation ATT,GTT I41V XP_016866236.1
XM_017010748.1 267 Intron XP_016866237.1

View Full Product Details