Product Details

SNP ID
rs181359494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:43866821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGAGGAAGTAGTACTTGTGTGGAG[A/T]CAACTGCCTCAGGACCACGGCACAG
Phenotype
MIM: 611986 MIM: 610337
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MRPS24 PubMed Links

Gene Details

Gene
MRPS24
Gene Name
mitochondrial ribosomal protein S24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032014.2 433 Missense Mutation ACT,TCT T128S NP_114403.1
Gene
URGCP
Gene Name
upregulator of cell proliferation
There are no transcripts associated with this gene.

Gene
URGCP-MRPS24
Gene Name
URGCP-MRPS24 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204871.1 433 UTR 3 NP_001191800.1

View Full Product Details