Product Details

SNP ID
rs181619657
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:60426808 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTACACCAAGAGCAGCCAGCCGGC[C/T]AAGTTCCTGGTGAGGCGCAAGACAT
Phenotype
MIM: 607740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C17orf64 PubMed Links

Gene Details

Gene
C17orf64
Gene Name
chromosome 17 open reading frame 64
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181707.2 805 Silent Mutation GCC,GCT A140A NP_859058.2
XM_005257033.4 805 Silent Mutation GCC,GCT A173A XP_005257090.1
XM_005257034.4 805 Silent Mutation GCC,GCT A173A XP_005257091.1
XM_005257035.4 805 Silent Mutation GCC,GCT A103A XP_005257092.1
XM_005257036.2 805 Silent Mutation GCC,GCT A140A XP_005257093.1
XM_017024166.1 805 Silent Mutation GCC,GCT A103A XP_016879655.1
Gene
RPL12P38
Gene Name
ribosomal protein L12 pseudogene 38
There are no transcripts associated with this gene.

Gene
USP32
Gene Name
ubiquitin specific peptidase 32
There are no transcripts associated with this gene.

View Full Product Details