Product Details

SNP ID
rs181755164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:102126904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATAGACAATGAAGAACAGCCAC[C/T]GCACGAGGGAAAGCCAGAAGTAGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TCEAL2 PubMed Links

Gene Details

Gene
TCEAL2
Gene Name
transcription elongation factor A like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080390.3 293 Missense Mutation CCG,CTG P25L NP_525129.1

View Full Product Details