Product Details
- SNP ID
-
rs181424547
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:31303265 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTAACTGACAAGTTGGAGTTAGAGT[A/C]GTAATCCTCCTTCACATTTTTAGAA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB116
PubMed Links
Gene Details
- Gene
- DEFB116
- Gene Name
- defensin beta 116
View Full Product Details