Product Details

SNP ID
rs181484364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19630531 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCTCTCTGGTCACTCTGTGGAG[C/T]TGGAATCTGCAGGGAGAAACCCAAG
Phenotype
MIM: 609694 MIM: 605110
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GMIP PubMed Links

Gene Details

Gene
GMIP
Gene Name
GEM interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288998.1 2530 Missense Mutation ACT,GCT T798A NP_001275927.1
NM_001288999.1 2530 Missense Mutation ACT,GCT T801A NP_001275928.1
NM_016573.3 2530 Missense Mutation ACT,GCT T827A NP_057657.2
XM_005259927.2 2530 Missense Mutation ACT,GCT T824A XP_005259984.1
XM_011528048.2 2530 Missense Mutation ACT,GCT T706A XP_011526350.1
XM_017026861.1 2530 Missense Mutation ACT,GCT T827A XP_016882350.1
XM_017026862.1 2530 Intron XP_016882351.1
Gene
LPAR2
Gene Name
lysophosphatidic acid receptor 2
There are no transcripts associated with this gene.

View Full Product Details