Product Details

SNP ID
rs181533598
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59477991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGTCCACCGGGTCCTCAAACTG[A/G]CCCATACAGACATTTTCATACTTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4D10 PubMed Links

Gene Details

Gene
OR4D10
Gene Name
olfactory receptor family 4 subfamily D member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004705.1 562 Missense Mutation ACC,GCC T188A NP_001004705.1

View Full Product Details