Product Details

SNP ID
rs181554393
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64895335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGGCCACACCCTCCCGCAGGTC[A/G]GCAGGCTGCTGGCCCCTGCGCAGCC
Phenotype
MIM: 616225 MIM: 610939 MIM: 610941
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATG2A PubMed Links

Gene Details

Gene
ATG2A
Gene Name
autophagy related 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015104.2 3261 Silent Mutation GCC,GCT A1845A NP_055919.2
XM_005273849.3 3261 Silent Mutation GCC,GCT A1839A XP_005273906.1
XM_005273850.3 3261 Silent Mutation GCC,GCT A1837A XP_005273907.1
XM_011544863.2 3261 Silent Mutation GCC,GCT A1847A XP_011543165.1
XM_011544864.2 3261 Silent Mutation GCC,GCT A1842A XP_011543166.1
XM_011544865.2 3261 Silent Mutation GCC,GCT A1782A XP_011543167.1
XM_011544866.2 3261 Intron XP_011543168.1
XM_011544867.2 3261 Intron XP_011543169.1
XM_011544868.2 3261 Silent Mutation GCC,GCT A1059A XP_011543170.1
Gene
MIR192
Gene Name
microRNA 192
There are no transcripts associated with this gene.

Gene
MIR194-2
Gene Name
microRNA 194-2
There are no transcripts associated with this gene.

Gene
MIR194-2HG
Gene Name
MIR194-2 host gene
There are no transcripts associated with this gene.

Gene
MIR6749
Gene Name
microRNA 6749
There are no transcripts associated with this gene.

Gene
MIR6750
Gene Name
microRNA 6750
There are no transcripts associated with this gene.

View Full Product Details