Product Details
- SNP ID
-
rs182054421
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:9757773 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTCTTAAATGGGTACTAAGGCCC[A/G]AGGATTGAGTGAAAGCTTTCCCACA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF846
PubMed Links
Gene Details
- Gene
- ZNF846
- Gene Name
- zinc finger protein 846
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001077624.1 |
2275 |
Missense Mutation |
TCG,TTG |
S435L |
NP_001071092.1 |
XM_005259772.3 |
2275 |
Missense Mutation |
TCG,TTG |
S435L |
XP_005259829.1 |
XM_005259773.4 |
2275 |
Missense Mutation |
TCG,TTG |
S434L |
XP_005259830.1 |
XM_005259775.3 |
2275 |
Missense Mutation |
TCG,TTG |
S434L |
XP_005259832.1 |
XM_006722658.3 |
2275 |
Intron |
|
|
XP_006722721.1 |
XM_011527717.2 |
2275 |
Missense Mutation |
TCG,TTG |
S435L |
XP_011526019.1 |
XM_011527718.2 |
2275 |
Missense Mutation |
TCG,TTG |
S434L |
XP_011526020.1 |
XM_011527719.2 |
2275 |
Missense Mutation |
TCG,TTG |
S399L |
XP_011526021.1 |
XM_011527720.2 |
2275 |
Missense Mutation |
TCG,TTG |
S399L |
XP_011526022.1 |
XM_011527721.2 |
2275 |
Missense Mutation |
TCG,TTG |
S378L |
XP_011526023.1 |
XM_017026404.1 |
2275 |
Missense Mutation |
TCG,TTG |
S421L |
XP_016881893.1 |
XM_017026405.1 |
2275 |
Missense Mutation |
TCG,TTG |
S306L |
XP_016881894.1 |
XM_017026406.1 |
2275 |
Missense Mutation |
TCG,TTG |
S306L |
XP_016881895.1 |
XM_017026407.1 |
2275 |
Intron |
|
|
XP_016881896.1 |
XM_017026408.1 |
2275 |
Intron |
|
|
XP_016881897.1 |
View Full Product Details